EMAIL THIS PAGE TO A FRIEND
Microvascular Research
Deletion of prolyl hydroxylase domain proteins (PHD1, PHD3) stabilizes hypoxia inducible factor-1 alpha, promotes neovascularization, and improves perfusion in a murine model of hind-limb ischemia.
PMID 25446011