EMAIL THIS PAGE TO A FRIEND
Journal of vascular surgery
Ephrin type-B receptor 4 activation reduces neointimal hyperplasia in human saphenous vein in vitro.
PMID 25446283
Abstract
Vein bypass is an essential therapy for patients with advanced peripheral and coronary artery disease despite development of neointimal hyperplasia. We have shown that stimulation of the receptor tyrosine kinase ephrin type-B receptor 4 (Eph-B4) with its ligand ephrin-B2 prevents neointimal hyperplasia in murine vein grafts. This study determines whether Eph-B4 in adult human veins is capable of phosphorylation and activation of downstream signaling pathways, as well as functional to release nitric oxide (NO) and prevent neointimal hyperplasia in vitro. Discarded human saphenous veins were taken from the operating room and placed in organ culture without or with ephrin-B2/Fc (2 μg/mL) for 14 days, and the neointima/media ratio was measured in matched veins. Primary human umbilical vein endothelial cells were treated with ephrin-B2/Fc (2 μg/mL) and examined with quantitative polymerase chain reaction, Western blot, immunoassays, and for release of NO. Ephrin-B2/Fc (2 μg/mL) was placed on the adventitia of saphenous veins treated with arterial shear stress for 24 hours in a bioreactor and activated Eph-B4 examined with immunofluorescence. The baseline intima/media ratio in saphenous vein rings was 0.456 ± 0.097, which increased to 0.726 ± 0.142 in untreated veins after 14 days in organ culture but only to 0.630 ± 0.132 in veins treated with ephrin-B2/Fc (n = 19, P = .017). Ephrin-B2/Fc stimulated Akt, endothelial NO synthase and caveolin-1 phosphorylation, and NO release (P = .007) from human umbilical vein endothelial cells (n = 6). Ephrin-B2/Fc delivered to the adventitia stimulated endothelial Eph-B4 phosphorylation after 24 hours of arterial stress in a bioreactor (n = 3). Eph-B4 is present and functional in adult human saphenous veins, with intact downstream signaling pathways capable of NO release and prevention of neointimal hyperplasia in vitro. Adventitial delivery of ephrin-B2/Fc activates endothelial Eph-B4 in saphenous veins treated with arterial shear stress in vitro. These results suggest that stimulation of Eph-B4 function may be a candidate strategy for translation to human clinical trials designed to inhibit venous neointimal hyperplasia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9405
O-Phospho-L-tyrosine
C9H12NO6P