EMAIL THIS PAGE TO A FRIEND
Environmental monitoring and assessment
Elevated titanium levels in Iraqi children with neurodevelopmental disorders echo findings in occupation soldiers.
PMID 25446717
Abstract
Anthropogenic release of pollutants into the environment is especially harmful to growing fetuses and young children. These populations are at an increased risk of damage because exposure to pollutants during critical periods of development can cause many impairments. Children's exposure to mixtures of metals could be responsible for the rising numbers of neurological disorders surfacing in Iraqi children. Titanium (Ti) and magnesium (Mg) are heavily used in war industries. Exposure to Ti and Mg has been linked to the dust in occupation soldiers' lungs. Hair samples of children in Hawija, Iraq (n = 13) contained significantly higher levels of Ti compared to Iranian children (n = 13) living near the Iraqi border (2080 ± 940 vs 707 ± 421 μg/kg, p < 0.0001). Magnesium was 1.7 times higher in Hawija children compared to Iranian children (115,763 ± 118,155 vs 67,650 ± 46,729 μg/kg). In samples from Hawija, Ti was 1.3 times higher in children with neurodevelopmental disorders (2198 ± 1108 vs 1942 ± 779 μg/kg), and Mg was 1.9 times higher in children without neurodevelopmental disorders (155,618 ± 140,791 vs 81,602 ± 91,940 μg/kg). Lead, arsenic, and cadmium in Hawija children with neurodevelopmental disorders (n = 6) were 2.5, 2.2, and 1.37 times higher compared to non-disabled children (n = 7). To get a clear understanding of the current status of neurodevelopmental disorders in Iraqi children and to determine the magnitude of this suspected global health issue, registries should be set up to compile and aggregate data from hospitals, clinics, and health centers across the country. Functional registries can develop collaborations with researchers toward finding causes of these disorders in Iraqi children and toward preventing them.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
366994
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Ti
267503
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Ti
348791
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Ti
268496
Titanium, powder, −100 mesh, 99.7% trace metals basis
Ti
305812
Titanium, crystalline, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Ti
369489
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Ti
268526
Titanium, sponge, 3-19 mm, 99.5% trace metals basis
Ti
348848
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Ti
348864
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Ti
266035
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Ti
267481
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Ti
433667
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Ti
460400
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Ti
460397
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Ti
513415
Titanium, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 98.5% trace metals basis
Ti
267902
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Ti
266019
Titanium, wire, diam. 0.127 mm, 99.99% trace metals basis
Ti
GF03980932
Titanium, mesh, 150x150mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Ti
GF16031843
Titanium, mesh, 100x100mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Ti
348805
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Ti
GF11328252
Titanium, mesh, 50x50mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Ti
348856
Titanium, wire, diam. 2.0 mm, 99.99% trace metals basis
Ti
767506
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Ti
GF71783354
Titanium, wire reel, 100m, diameter 0.75mm, as drawn, 99.6+%
Ti
347132
Titanium, rod, diam. 6.35 mm, 99.99% trace metals basis
Ti
GF76735963
Titanium, wire reel, 10m, diameter 0.75mm, as drawn, 99.6+%
Ti
GF29374005
Titanium, mesh, 100x100mm, nominal aperture 4.3mm, wire diameter 1.5mm, 4.4x4.4 wires/inch, open area 94%, platinized diamond mesh
Ti
GF34739837
Titanium, sphere, 200pcs, diameter 3mm, 99.6+%
Ti
IRMM531A
Titanium, IRMM® certified Reference Material, 0.1 mm foil
Ti
GF64061760
Titanium, rod, 1000mm, diameter 16mm, annealed, 99.6+%
Ti