EMAIL THIS PAGE TO A FRIEND
Journal of Renal Nutrition : the Official Journal of the Council on Renal Nutrition of the National Kidney Foundation
Fiber supplementation lowers plasma p-cresol in chronic kidney disease patients.
PMID 25446837