EMAIL THIS PAGE TO A FRIEND
European journal of immunology
Differential expression of the immunosuppressive enzyme IL4I1 in human induced Aiolos+, but not natural Helios+, FOXP3+ Treg cells.
PMID 25446972
Abstract
IL4I1 encodes an L-phenylalanine oxidase that inhibits T-cell proliferation. It has been recently reported that IL4I1 is expressed in TH17 cells as part of a mechanism that limits their pathogenicity. We have previously identified a population of human FOXP3(+) Treg cells that secrete IL-17 ex vivo; here, we addressed the expression of IL4I1 in that Treg-cell population. We found that in ex vivo isolated circulating Treg cells, IL4I1 expression is induced by activation. Moreover, IL4I1 expression is restricted to cells that do not express Helios, a transcription factor that characterizes natural Treg cells, but that express Aiolos, which is involved in the differentiation of TH17 and induced Treg cells. We also showed that conversion of Treg cells under inflammatory conditions increases IL4I1 expression, likely as part of a regulatory loop that attempts to limit the pathogenicity resulting from their conversion into TH17. The specific expression of IL4I1 in TH17 and iTreg cells may provide insights into approaches that aim at modulating these populations in different pathological conditions involving inflammation-mediated immunosuppression.