EMAIL THIS PAGE TO A FRIEND
Experimental Cell Research
OSBP-related protein 3 (ORP3) coupling with VAMP-associated protein A regulates R-Ras activity.
PMID 25447204
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3766
Phorbol 12-myristate 13-acetate 4-O-methyl ether
C37H58O8