EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
The contribution of activating transcription factor 3 to apoptosis of human colorectal cancer cells by protocatechualdehyde, a naturally occurring phenolic compound.
PMID 25447816
Abstract
Protocatechualdehyde (PCA) is one of the important compounds found in barley, green cavendish bananas and grapevine leaves. PCA shows anti-cancer activities in breast, leukemia and colorectal cancer cells. Previous study reported that PCA exerts anti-cancer activity through down-regulating cyclin D1 and HDAC2 in human colorectal cancer cells. However, the underlying mechanisms for the expression of activating transcription factor 3 (ATF3) by PCA has not been studied. Thus, we performed in vitro study to investigate if treatment of PCA affects ATF3 expression and ATF3-mediated apoptosis in human colorectal cancer cells. PCA decreased cell viability in a dose-dependent manner in HCT116 and SW480 cells. In addition, PCA reduced cell viability in MCF-7, MDA-MB-231 and HepG-2 cells. Exposure of PCA activated the levels of ATF3 protein and mRNA in HCT116 and SW480 cells. Inhibition of ERK1/2/ by PD98059 and p38 by SB203580 inhibited PCA-induced ATF3 expression and transcriptional activation. ATF3-knockdown inhibited PCA-induced apoptosis and cell viability. In addition, ATF3 overexpression enhanced PCA-mediated cleavage of PARP. These findings suggest that inhibition of cell viability and apoptosis by PCA may be result of ATF3 expression through ERK1/2 and p38-mediated transcriptional activation.