EMAIL THIS PAGE TO A FRIEND
Experimental Neurology
Gene expression profiling studies in regenerating nerves in a mouse model for CMT1X: uninjured Cx32-knockout peripheral nerves display expression profile of injured wild type nerves.
PMID 25447941