EMAIL THIS PAGE TO A FRIEND
European Journal of Pharmaceutics and Biopharmaceutics
Ion milling coupled field emission scanning electron microscopy reveals current misunderstanding of morphology of polymeric nanoparticles.
PMID 25448070