EMAIL THIS PAGE TO A FRIEND
Acta biomaterialia
Functional fabrication of recombinant human collagen-phosphorylcholine hydrogels for regenerative medicine applications.
PMID 25448347
Abstract
The implant-host interface is a critical element in guiding tissue or organ regeneration. We previously developed hydrogels comprising interpenetrating networks of recombinant human collagen type III and 2-methacryloyloxyethyl phosphorylcholine (RHCIII-MPC) as substitutes for the corneal extracellular matrix that promote endogenous regeneration of corneal tissue. To render them functional for clinical application, we have now optimized their composition and thereby enhanced their mechanical properties. We have demonstrated that such optimized RHCIII-MPC hydrogels are suitable for precision femtosecond laser cutting to produce complementing implants and host surgical beds for subsequent tissue welding. This avoids the tissue damage and inflammation associated with manual surgical techniques, thereby leading to more efficient healing. Although we previously demonstrated in clinical testing that RHCIII-based implants stimulated cornea regeneration in patients, the rate of epithelial cell coverage of the implants needs improvement, e.g. modification of the implant surface. We now show that our 500μm thick RHCIII-MPC constructs comprising over 85% water are suitable for microcontact printing with fibronectin. The resulting fibronectin micropatterns promote cell adhesion, unlike the bare RHCIII-MPC hydrogel. Interestingly, a pattern of 30μm wide fibronectin stripes enhanced cell attachment and showed the highest mitotic rates, an effect that potentially can be utilized for faster integration of the implant. We have therefore shown that laboratory-produced mimics of naturally occurring collagen and phospholipids can be fabricated into robust hydrogels that can be laser profiled and patterned to enhance their potential function as artificial substitutes of donor human corneas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
02562
1,2-Dichloroethane, analytical standard
C2H4Cl2
A5508
Adenosine 5′-phosphosulfate sodium salt, ≥85%
C10H14N5O10PS
A3678
Ammonium persulfate, for molecular biology, for electrophoresis, ≥98%
H8N2O8S2
248614
Ammonium persulfate, ACS reagent, ≥98.0%
H8N2O8S2
215589
Ammonium persulfate, reagent grade, 98%
H8N2O8S2
A7460
Ammonium persulfate, BioXtra, ≥98.0%
H8N2O8S2
09913
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
H8N2O8S2
31117
Ammonium persulfate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
H8N2O8S2
C1359
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
C46H46N2O23
17783
Calcein-AM, BioReagent, suitable for fluorescence, ≥96.0% (HPLC)
C46H46N2O23
56496
Calcein-AM, Small Package (20 X 50 μg ), ≥96.0% (HPLC)
C46H46N2O23
M2933
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
C6H13NO4S · xH2O
M5287
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
C6H13NO4S · xH2O
69890
MES hydrate, BioUltra, ≥99.5% (T)
C6H13NO4S · xH2O
M8250
MES hydrate, ≥99.5% (titration)
C6H13NO4S · xH2O
PHG0003
MES hydrate, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C6H13NO4S · xH2O
69889
MES monohydrate, BioUltra, for molecular biology, ≥99.5% (T)
C6H13NO4S · H2O
69892
MES monohydrate, BioXtra, ≥99.0% (T)
C6H13NO4S · H2O
RES0113M-A7
MES monohydrate, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C6H13NO4S · H2O
T9281
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ~99%
C6H16N2
T7024
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
C6H16N2
411019
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
C6H16N2
T22500
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
C6H16N2
56480
N-Hydroxysuccinimide, purum, ≥97.0% (T)
C4H5NO3
235709
Octamethyltrisiloxane, 98%
C8H24O2Si3