EMAIL THIS PAGE TO A FRIEND
Photodiagnosis and Photodynamic Therapy
Polymeric micelles for enhanced Photofrin II ? delivery, cytotoxicity and pro-apoptotic activity in human breast and ovarian cancer cells.
PMID 25449154