EMAIL THIS PAGE TO A FRIEND
Biochimica et Biophysica Acta
Transglutaminase 2 interacts with syndecan-4 and CD44 at the surface of human macrophages to promote removal of apoptotic cells.
PMID 25449226