EMAIL THIS PAGE TO A FRIEND
Journal of Ethnopharmacology
Neuroprotective effect of modified Chungsimyeolda-tang, a traditional Korean herbal formula, via autophagy induction in models of Parkinson's disease.
PMID 25449460