EMAIL THIS PAGE TO A FRIEND
Comparative Biochemistry and Physiology. Part A, Molecular & Integrative Physiology
Nutritional stress in Northern gannets during an unprecedented low reproductive success year: can extreme sea surface temperature event and dietary change be the cause?
PMID 25449633