EMAIL THIS PAGE TO A FRIEND
Toxicology
Developmental exposure to purity-controlled polychlorinated biphenyl congeners (PCB74 and PCB95) in rats: effects on brainstem auditory evoked potentials and catalepsy.
PMID 25449634