EMAIL THIS PAGE TO A FRIEND
Gastroenterology
Systemic ulcerative lesions in a patient with ischemic heart disease.
PMID 25450090
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3539
Nicorandil, ≥98% (HPLC)
C8H9N3O4