EMAIL THIS PAGE TO A FRIEND
Journal of dermatological science
Decreased ATP synthesis and lower pH may lead to abnormal muscle contraction and skin sensitivity in human skin.
PMID 25450093
Abstract
Sensitive skin represents hyperactive sensory symptoms showing exaggerated reactions in response to internal stimulants or external irritants. Although sensitive skin is a very common condition affecting an estimated 50% of the population, its pathophysiology remains largely elusive, particularly with regard to its metabolic aspects. The objective of our study was to investigate the pathogenesis of sensitive skin. We recruited healthy participants with 'sensitive' or 'non-sensitive' skin based on standardized questionnaires and 10% lactic acid stinging test, and obtained skin samples for microarray analysis and subsequent experiments. Microarray transcriptome profiling revealed that genes involved in muscle contraction, carbohydrate and lipid metabolism, and ion transport and balance were significantly decreased in sensitive skin. These altered genes could account for the abnormal muscle contraction, decreased ATP amount in sensitive skin. In addition, pain-related transcripts such as TRPV1, ASIC3 and CGRP were significantly up-regulated in sensitive skin, compared with non-sensitive skin. Our findings suggest that sensitive skin is closely associated with the dysfunction of muscle contraction and metabolic homeostasis.