EMAIL THIS PAGE TO A FRIEND
Biochimica et Biophysica Acta
Phospholipid lateral diffusion in phosphatidylcholine-sphingomyelin-cholesterol monolayers; effects of oxidatively truncated phosphatidylcholines.
PMID 25450344