EMAIL THIS PAGE TO A FRIEND
Biochimica et Biophysica Acta
Endothelial Nlrp3 inflammasome activation associated with lysosomal destabilization during coronary arteritis.
PMID 25450976
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2580000
Colchicine, (European Pharmacopoeia (EP) Reference Standard)
C22H25NO6