EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
Orientation of myosin binding protein C in the cardiac muscle sarcomere determined by domain-specific immuno-EM.
PMID 25451032
Abstract
Myosin binding protein C is a thick filament protein of vertebrate striated muscle. The cardiac isoform [cardiac myosin binding protein C (cMyBP-C)] is essential for normal cardiac function, and mutations in cMyBP-C cause cardiac muscle disease. The rod-shaped molecule is composed primarily of 11 immunoglobulin- or fibronectin-like domains and is located at nine sites, 43nm apart, in each half of the A-band. To understand how cMyBP-C functions, it is important to know its structural organization in the sarcomere, as this will affect its ability to interact with other sarcomeric proteins. Several models, in which cMyBP-C wraps around, extends radially from, or runs axially along the thick filament, have been proposed. Our goal was to define cMyBP-C orientation by determining the relative axial positions of different cMyBP-C domains. Immuno-electron microscopy was performed using mouse cardiac myofibrils labeled with antibodies specific to the N- and C-terminal domains and to the middle of cMyBP-C. Antibodies to all regions of the molecule, except the C-terminus, labeled at the same nine axial positions in each half A-band, consistent with a circumferential and/or radial rather than an axial orientation of the bulk of the molecule. The C-terminal antibody stripes were slightly displaced axially, demonstrating an axial orientation of the C-terminal three domains, with the C-terminus closer to the M-line. These results, combined with previous studies, suggest that the C-terminal domains of cMyBP-C run along the thick filament surface, while the N-terminus extends toward neighboring thin filaments. This organization provides a structural framework for understanding cMyBP-C's modulation of cardiac muscle contraction.