EMAIL THIS PAGE TO A FRIEND
Journal of Reproduction and Development
Potential role of hCG in apoptosis of human luteinized granulosa cells.
PMID 25451535
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000005 Amphotericin B, European Pharmacopoeia (EP) Reference Standard
C47H73NO17
A9528
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
C47H73NO17
D8900 Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
76524
Glycine, certified reference material, TraceCERT®
C2H5NO2
50049
Glycine, BioUltra, ≥99.0% (NT)
C2H5NO2
50050
Glycine, ≥99.0% (NT)
C2H5NO2
50058
Glycine, tested according to Ph.Eur.
C2H5NO2
PHR1111
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
C2H5NO2
1295800
Glycine, United States Pharmacopeia (USP) Reference Standard
C2H5NO2
94119
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
C2H5NO2
G0450000
Glycine, European Pharmacopoeia (EP) Reference Standard
C2H5NO2
82762
Methanol, analytical standard
CH4O
PHR1372
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
CH4O
1424109
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
CH4O
M8046
Mifepristone, ≥98%
C29H35NO2
P3532
Phenol Red, powder, BioReagent, suitable for cell culture
C19H14O5S
114529
Phenol Red, ACS reagent
C19H14O5S
S9133
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Na2O3Se
S5261
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Na2O3Se
71950
Sodium selenite, anhydrous, ≥95.0% (RT)
Na2O3Se
214485
Sodium selenite, 99%
Na2O3Se