EMAIL THIS PAGE TO A FRIEND
Prostaglandins, Leukotrienes, and Essential Fatty Acids
A method for long term stabilisation of long chain polyunsaturated fatty acids in dried blood spots and its clinical application.
PMID 25451557