EMAIL THIS PAGE TO A FRIEND
Journal of cardiac failure
Novel extracellular matrix biomarkers as predictors of adverse outcome in chronic heart failure: association between biglycan and response to statin therapy in the CORONA trial.
PMID 25451704
Abstract
The extracellular matrix (ECM) plays an important role in left ventricular remodeling and progression of heart failure (HF). Biglycan and mimecan are ECM proteins that are abundantly expressed in cardiac tissue but have not been evaluated as prognostic markers in HF. We investigated their interaction with statin treatment and association with adverse outcome in chronic HF. The association between serum levels of biglycan and mimecan and the primary end point (cardiovascular [CV] death, nonfatal myocardial infarction, nonfatal stroke), all-cause mortality, CV death, the composite of all-cause mortality/hospitalization for worsening of HF, and the coronary end point was evaluated in 1,390 patients >60 years of age with ischemic systolic HF in the Controlled Rosuvastatin Multinational Trial in HF (CORONA) population, randomly assigned to 10 mg rosuvastatin or placebo. Serum biglycan and mimecan added no prognostic information beyond conventional risk factors, including N-terminal pro-B-type natriuretic peptide. However, statin treatment improved all outcomes except CV death in patients with low biglycan levels (ie, lower tertile), even after full multivariable adjustment. Although circulating levels of mimecan and biglycan were of limited predictive value in patients with chronic HF, circulating biglycan could be a useful marker for targeting statin therapy in patients with HF.