EMAIL THIS PAGE TO A FRIEND
Neurotoxicology
Neuroprotective role of prostaglandin PGE2 EP2 receptor in hemin-mediated toxicity.
PMID 25451967
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51280
Hemin, BioXtra, from Porcine, ≥97.0% (HPLC)
C34H32ClFeN4O4
H9039
Hemin, from bovine, ≥90%
C34H32ClFeN4O4