EMAIL THIS PAGE TO A FRIEND
Gene
Reconstruction and analysis of the genome-scale metabolic model of Lactobacillus casei LC2W.
PMID 25452194
Abstract
Lactobacillus casei LC2W is a recently isolated probiotic lactic acid bacterial strain, which is widely used in the dairy and pharmaceutical industries and in clinical medicine. The first genome-scale metabolic model for L. casei, composed of 846 genes, 969 metabolic reactions, and 785 metabolites, was reconstructed using both manual genome annotation and an automatic SEED model. Then, the iJL846 model was validated by simulating cell growth on 15 reported carbon sources. The iJL846 model explored the metabolism of L. casei on a genome scale: (1) explanation of the genetic codes-metabolic functions of 342 genes were reannotated in this model; (2) characterization of the physiology-10 amino acids and 7 vitamins were identified to be essential nutrients for L. casei LC2W growth; (3) analyses of metabolic pathways-the transport and metabolism of the 17 essential nutrients and exopolysaccharide (EPS) biosynthesis-were performed; (4) exploration of metabolic capacity was conducted-for lactate, the importance of genes in its biosynthetic pathways was evaluated, and the requirements of amino acids were predicted for mixed acid fermentation; for flavor compounds, the effects of oxygen were analyzed, and three new knockout targets were selected for acetoin production; for EPS, 11 types of nutrients in the rich medium and important reactions in the biosynthetic pathway were identified that enhanced EPS production. In conclusion, the iJL846 model serves as a useful tool for understanding and engineering the metabolism of this probiotic strain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2764
Activated charcoal, untreated, granular, ≤5 mm
C
C2889
Activated charcoal, untreated, granular, 8-20 mesh
C
161551
Activated charcoal, powder, -100 particle size (mesh), decolorizing
C
292591
Activated charcoal, 4-14 mesh, granular, Norit® PK 3-5
C
242233
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
C
242241
Activated charcoal, DARCO®, 12-20 mesh, granular
C
242268
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
C
242276
Activated charcoal, DARCO®, −100 mesh particle size, powder
C
329428
Activated charcoal, NORIT® ROW 0.8 SUPRA, pellets
C
C3014
Activated charcoal, untreated, granular, 20-60 mesh
C
C4386
Activated charcoal, acid-washed with hydrochloric acid
C
C5510
Activated charcoal, acid-washed with phosphoric and sulfuric acids
C
05112
Activated charcoal, for GC
C
675326
Activated charcoal, DARCO® KB-G
C
80485
Activated charcoal, for the determination of AOX, 50-150 μm particle size
C
C7606
Activated Charcoal, meets USP testing specifications
C
97876
Activated Charcoal Norit®, Norit CA1, from wood, chemically activated, powder
C
37771
Activated Charcoal Norit®, Norit GAC 1240W, from coal, for potable water processing, steam activated, granular
C
22874
Activated Charcoal Norit®, Norit PK 1-3, from peat, steam activated, granular
C
29204
Activated Charcoal Norit®, Norit RB3, for gas purification, steam activated, rod
C
51127
Activated Charcoal Norit®, Norit RB4C, steam activated, rod, for the purification of gas
C
53663
Activated Charcoal Norit®, Norit SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
C
93067
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
C
484164 Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
C
633100 Carbon, nanopowder, <100 nm particle size (TEM)
C
GF29093058
Carbon, chopped fiber, 100g, nominal diameter 0.007mm, fiber length 6mm, grade 34-700
C
GF02887342
Carbon, fabric, 1000x1000mm, weight.muü 375g, thickness 0.4mm, ends x picks/10cm 47x47, 2/2 twill weave
C
GF33494957
Carbon, fabric, 1000x1000mm, weight.muü 92g, thickness 0.15mm, ends x picks/10cm 69x69, plain weave
C
GF07091861
Carbon, fabric, 150x150mm, weight.muü 375g, thickness 0.4mm, ends x picks/10cm 47x47, 2/2 twill weave
C
GF39337724
Carbon, fabric, 250x250mm, weight.muü 92g, thickness 0.15mm, ends x picks/10cm 69x69, plain weave
C