EMAIL THIS PAGE TO A FRIEND
Infection and Immunity
Klebsiella pneumoniae translocates across the intestinal epithelium via Rho GTPase- and phosphatidylinositol 3-kinase/Akt-dependent cell invasion.
PMID 25452552
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92136
Genistein, analytical standard
C15H10O5