EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Effect of kidney-reinforcing and marrow-beneficial traditional Chinese medicine-intervened serum on the proliferation and osteogenic differentiation of bone marrow stromal cells.
PMID 25452801
Abstract
The present study aimed to investigate the effect of kidney-reinforcing and marrow-beneficial traditional Chinese medicine (TCM)-intervened (KRMBTI)-serum on the proliferation and osteogenic differentiation of bone marrow stromal cells (BMSCs) in rats. Rat BMSCs were isolated and cultured