EMAIL THIS PAGE TO A FRIEND
Biomaterials
Endochondral bone formation in gelatin methacrylamide hydrogel with embedded cartilage-derived matrix particles.
PMID 25453948
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
155721
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
C4H6O2