EMAIL THIS PAGE TO A FRIEND
Biomaterials
Novel protease inhibitor-loaded Nanoparticle-in-Microparticle Delivery System leads to a dramatic improvement of the oral pharmacokinetics in dogs.
PMID 25453966