EMAIL THIS PAGE TO A FRIEND
Chemistry and Physics of Lipids
NMR-based molecular ruler for determining the depth of intercalants within the lipid bilayer. Part V: a comparison of liposomes, bioliposomes and erythrocyte ghosts.
PMID 25454361