EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Biomonitoring occupational sevoflurane exposure at low levels by urinary sevoflurane and hexafluoroisopropanol.
PMID 25455444
Abstract
This study aimed to correlate environmental sevoflurane levels with urinary concentrations of sevoflurane (Sev-U) or its metabolite hexafluoroisopropanol (HFIP) in order to assess and discuss the main issues relating to which biomarker of sevoflurane exposure is best, and possibly suggest the corresponding biological equivalent exposure limit values. Individual sevoflurane exposure was measured in 100 healthcare operators at five hospitals in north-east Italy using the passive air sampling device Radiello(®), and assaying Sev-U and HFIP concentrations in their urine collected at the end of the operating room session. All analyses were performed by gas chromatography-mass spectrometry. Environmental sevoflurane levels in the operating rooms were also monitored continuously using an infrared photoacoustic analyzer. Our results showed very low individual sevoflurane exposure levels, generally below 0.5 ppm (mean 0.116 ppm; range 0.007-0.940 ppm). Sev-U and HFIP concentrations were in the range of 0.1-17.28 μg/L and 5-550 μg/L, respectively. Both biomarkers showed a statistically significant correlation with the environmental exposure levels (Sev-U, r=0.49; HFIP, r=0.52), albeit showing fairly scattered values. Sev-U values seem to be influenced by peaks of exposure, especially at the end of the operating-room session, whereas HFIP levels by exposure on the previous day, the data being consistent with the biomarkers' very different half-lives (2.8 and 19 h, respectively). According to our results, both Sev-U and HFIP are appropriate biomarkers for assessing sevoflurane exposure at low levels, although with some differences in times/patterns of exposure. More work is needed to identify the best biomarker of sevoflurane exposure and the corresponding biological equivalent exposure limit values.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1462
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
C4H7N3O
RAD141S
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
RAD125
radiello Anesthetic Gases and Vapor Sampler
RAD190
radiello Barcode Labels, pk of 198
RAD405
radiello BTEX Calibration Kit (CS2 Desorption)
RAD407 radiello BTEX Calibration Kit (Thermal Desorption)
RAD130S
radiello BTEX/VOC Starter Kit, CS2 Desorption
RAD145S
radiello BTEX/VOC Starter Kit, Thermal Desorption
RAD165
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
RAD168
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
RAD132
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
RAD130
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
RAD145
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
RAD195 radiello Clips, suspending Radiello triangular support plate, pk of 20
RAD120
radiello Diffusive Bodies, white, configured for general use, pk of 20
RAD1201
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
RAD1202
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
RAD1203
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
RAD1991
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
RAD1992
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
RAD174
radiello Filtration Kit, pk of 20
RAD170S
radiello Hydrogen Sulfide Starter Kit
RAD171 radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
RAD126
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
RAD1261
radiello On-Field Thermometer, configured for ready-to-use (rtu) samplers, Include one rtu vertical adapter for each thermometer, pk of 3
RAD127
radiello On-Field Thermometer, Reader, pk of 1
RAD196
radiello Outdoor Shelter, pk of 10
RAD198 radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
RAD121
radiello Triangular Support Plate, pk of 20
RAD122
radiello Vertical Adapter, threaded for standard use, pk of 20