EMAIL THIS PAGE TO A FRIEND
Food And Chemical Toxicology
Rosmarinic acid attenuates hepatic ischemia and reperfusion injury in rats.
PMID 25455894
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1000
1-Naphthol, ReagentPlus®, ≥99%
C10H8O
31097
1-Naphthol, PESTANAL®, analytical standard
C10H8O
N2780
1-Naphthol, BioXtra, ≥99%
C10H8O
33420
1-Naphthol, puriss. p.a., Reag. Ph. Eur., ≥99% (GC)
C10H8O
70440
1-Naphthol, puriss., for fluorescence, ≥99.0% (GC)
C10H8O
1102974
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
C19H42BrN
ARK2188
Hexadecyltrimethylammonium bromide, PharmaGrade, USP/NF, Manufactured under appropriate controls for pharma or biopharmaceutical production.
C19H42BrN
H9151
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
C19H42BrN
52365
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
C19H42BrN
52369
Hexadecyltrimethylammonium bromide, BioUltra, ≥99.0% (AT)
C19H42BrN
H6269
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
C19H42BrN
52367
Hexadecyltrimethylammonium bromide, for ion pair chromatography
C19H42BrN
H5882
Hexadecyltrimethylammonium bromide, ≥98%
C19H42BrN
36932
Hexadecyltrimethylammonium bromide, analytical standard
C19H42BrN
52370
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
C19H42BrN
07883
Hexadecyltrimethylammonium bromide, puriss. p.a., matrix substance for MALDI-MS
C19H42BrN
72586
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
NNaO2
72587
Nitrite ion standard solution, 0.01 M NO2-, for ion-selective electrodes
NNaO2
563218
Sodium nitrite, 99.999% trace metals basis
NNaO2
431605
Sodium nitrite, ≥99.99% trace metals basis
NNaO2
524379
Sodium nitrite, 99.5%, super free-flowing
NNaO2
67398
Sodium nitrite, analytical standard
NNaO2
71760
Sodium nitrite, purum p.a., ≥98.0% (RT)
NNaO2