EMAIL THIS PAGE TO A FRIEND
Cancer science
Immunohistochemical analysis of RTKs expression identified HER3 as a prognostic indicator of gastric cancer.
PMID 25455899
Abstract
Standard treatment in Japan for the 13th Japanese Gastric Cancer Association stage II/III advanced gastric cancer is postoperative adjuvant S-1 administration after curative surgery. High expression of receptor type tyrosine kinases (RTKs) has repeatedly represented poor prognosis for cancers. However it has not been demonstrated whether RTKs have prognostic relevance for stage II/III gastric cancer with standard treatment. Tumor tissues were obtained from 167 stage II/III advanced gastric cancer patients who underwent curative surgery and received postoperative S-1 chemotherapy from 2000 to 2010. Expression of the RTKs including EGFR, HER2, HER3, IGF-1R, and EphA2 was analyzed using immunohistochemistry (IHC). Analysis using a multivariate proportional hazard model identified the most significant RTKs that represented independent prognostic relevance. When tumor HER3 expression was classified into IHC 1+/2+ (nxa0=xa098) and IHC 0 (nxa0=xa069), the cumulative 5-year Relapse Free Survival (5y-RFS) was 56.5 and 82.9%, respectively (Pxa0=xa00.0034). Significant prognostic relevance was similarly confirmed for IGF-1R (Pxa0=xa00.014), and EGFR (Pxa0=xa00.030), but not for EphA2 or HER2 expression. Intriguingly, HER3 expression was closely correlated with IGF-1R (Pxa0