EMAIL THIS PAGE TO A FRIEND
Biochimie
Isolation and characterization of four medium-size disintegrins from the venoms of Central American viperid snakes of the genera Atropoides, Bothrops, Cerrophidion and Crotalus.
PMID 25457103