EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Development and validation of method for TH588 and TH287, potent MTH1 inhibitors and new anti-cancer agents, for pharmacokinetic studies in mice plasma.
PMID 25459754
Abstract
MTH1 is a protein that is required for cancer cell survival and is overexpressed in cancer cells. TH588 and TH287 are two new compounds that inhibit the MTH1 protein. The inhibitors were tested in pharmacokinetic studies on mice. A bioanalytical method was developed and validated for determination in mice plasma. The method was based on protein precipitation followed by LC-MS/MS analysis. The separation was performed on an Ascentis Express RP-Amide C18 column. The mass spectrometer was operated in positive electrospray ionization mode and the analytes were determined with multiple reaction monitoring (MRM). Abundant monoisotopic fragments were used for quantification. Two additional fragments were used for conformational analysis. The recovery of the compounds in plasma varied between 61 and 91% and the matrix effects were low and ranged between -3% and +2%. The method showed to be selective, linear, accurate and precise, and applicable for preclinical pharmacokinetic studies of TH588 and TH287 in mouse plasma. Half-life (T1/2) was ≤3.5h and maximum concentration (Cmax) ranged between 0.82 and 338μM for the different administration routes and compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45983
Acetonitrile, analytical standard
C2H3N
PHR1551
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
00712
Acetonitrile solution, ~20% in H2O, for protein sequence analysis
C2H3N
516961
Ammonium formate, ≥99.995% trace metals basis
CH5NO2
09735
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
CH5NO2
70221
Ammonium formate, for mass spectrometry, ≥99.0%
CH5NO2
78314
Ammonium formate solution, BioUltra, 10 M in H2O
CH5NO2
714690
Ammonium formate solution, 10 mM in H2O, for HPLC
CH5NO2
50461-U Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
50462-U Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
50822-U Ascentis® Express C18, 2 Micron Guard Cartridge, 2 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
50823-U Ascentis® Express C18, 2 Micron Guard Cartridge, 2 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
50805-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 2 cm × 2.1 mm
50809-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 2.1 mm
50811-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 2.1 mm
50812-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 2.1 mm
50813-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 2.1 mm
50814-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 2.1 mm
50815-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 3 mm
50816-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 3 mm
50817-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 3 mm
50819-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 3 mm
50821-U Ascentis® Express C18, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 3 mm
53982-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 75 μm
53985-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 100 μm
53989-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 200 μm
53992-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 300 μm
53998-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 500 μm
54219-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 75 μm
54256-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 100 μm