EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Bioassay-guided preparative separation of angiotensin-converting enzyme inhibitory C-flavone glycosides from Desmodium styracifolium by recycling complexation high-speed counter-current chromatography.
PMID 25459924
Abstract
A new strategy of the convergence of high-speed counter-current chromatography (HSCCC) and bioactive assay technique was developed for rapidly screening and separating the angiotensin-converting enzyme (ACE) inhibitors from the aerial parts of Desmodium styracifolium. Bioactivity-guided fractionation of the crude extract was first established to target the bioactive fractions based on HSCCC coupled with in vitro ACE inhibitory assay. Subsequently, the bioactive fractions were further separated by the recycling complexation HSCCC respectively, using 0.10 mol/L copper sulfate in the lower phase of two-phase solvent system composed of n-butanol/water (1:1, v/v). Five C-glycosylflavones, vicenin 2 (1), carlinoside (2), vicenin 1 (3), schaftoside (4) and vicenin 3 (5), were successfully obtained. Their chemical structures were identified using ESI-MS and NMR. All the isolates showed in vitro ACE inhibitory activity with the IC50 values between 33.62 and 58.37 μM. The results demonstrated that the established method was proposed as an excellent strategy to systematically screen and purify active compounds from traditional Chinese medicines.