EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Identification of hydrolytic and isomeric N-oxide degradants of vilazodone by on line LC-ESI-MS/MS and APCI-MS.
PMID 25459935