EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Plasma protein binding, pharmacokinetics, tissue distribution and CYP450 biotransformation studies of fidarestat by ultra high performance liquid chromatography-high resolution mass spectrometry.
PMID 25459938
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS019917
Adenine phosphate salt, AldrichCPR
C5H5N5 · H3O4P
A9272
Adenosine 3′-monophosphate, from yeast
C10H14N5O7P