EMAIL THIS PAGE TO A FRIEND
Environmental research
Renal and blood pressure effects from environmental cadmium exposure in Thai children.
PMID 25460624
Abstract
Very few studies have shown renal and blood pressure effects from environmental cadmium exposure in children. This population study examined associations between urinary cadmium excretion, a good biomarker of long-term cadmium exposure, and renal dysfunctions and blood pressure in environmentally exposed Thai children. Renal functions including urinary excretion of β2-microglobulin, calcium (early renal effects), and total protein (late renal effect), and blood pressure were measured in 594 primary school children. Of the children studied, 19.0% had urinary cadmium ≥ 1 μg/g creatinine. The prevalence of urinary cadmium ≥ 1 μg/g creatinine was significantly higher in girls and in those consuming rice grown in cadmium-contaminated areas. The geometric mean levels of urinary β2-microglobulin, calcium, and total protein significantly increased with increasing tertiles of urinary cadmium. The analysis did not show increased blood pressure with increasing tertiles of urinary cadmium. After adjusting for age, sex, and blood lead levels, the analysis showed significant positive associations between urinary cadmium and urinary β2-microglobulin and urinary calcium, but not urinary total protein nor blood pressure. Our findings provide evidence that environmental cadmium exposure can affect renal functions in children. A follow-up study is essential to assess the clinical significance and progress of renal effects in these children.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00623
Cadmium, purum p.a., for filling reductors
Cd
265411
Cadmium, foil, thickness 0.5 mm, ≥99.99% trace metals basis
Cd
265330
Cadmium, granular, ≥99%, 5-20 mesh
Cd
414891
Cadmium, granular, 30-80 mesh, ≥99%
Cd
265365
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Cd
265454
Cadmium, rod, diam. 4.0 mm, 99.999% trace metals basis
Cd
202886
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cd
348600
Cadmium, wire, diam. 1.0 mm, 99.999% trace metals basis
Cd
GF61936814
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF17721637
Cadmium, foil, 100x100mm, thickness 0.5mm, as rolled, 99.95%
Cd
GF64286602
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF30096345
Cadmium, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF10741889
Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF37649100
Cadmium, foil, 100x200mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF38583287
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF50990597
Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF72757503
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF66037754
Cadmium, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Cd
GF76768914
Cadmium, foil, 150x150mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF33512688
Cadmium, foil, 200x200mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF94010453
Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF90633652
Cadmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Cd
GF06836238
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cd
GF71408763
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF59435062
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cd
GF78263411
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cd
GF84554613
Cadmium, foil, 300x1000mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF19008817
Cadmium, foil, 300x300mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF20958947
Cadmium, foil, 300x300mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF15879954
Cadmium, foil, 300x300mm, thickness 2.0mm, as rolled, 99.99%
Cd