EMAIL THIS PAGE TO A FRIEND
Environmental Research
Toxicity of Bacillus thuringiensis var. israelensis in aqueous suspension on the South American common frog Leptodactylus latrans (Anura: Leptodactylidae) tadpoles.
PMID 25460638