EMAIL THIS PAGE TO A FRIEND
Environmental research
Genotoxic and cell-transforming effects of titanium dioxide nanoparticles.
PMID 25460650
Abstract
The in vitro genotoxic and the soft-agar anchorage independent cell transformation ability of titanium dioxide nanoparticles (nano-TiO2) and its microparticulated form has been evaluated in human embryonic kidney (HEK293) and in mouse embryonic fibroblast (NIH/3T3) cells. Nano-TiO2 of two different sizes (21 and 50 nm) were used in this study. The comet assay, with and without the use of FPG enzyme, the micronucleus assay and the soft-agar colony assay were used. For both the comet assay and the frequency of micronuclei a statistically significant induction of DNA damage, was observed at the highest dose tested (1000 µg/mL). No oxidative DNA damage induction was observed when the comet assay was complemented with the use of FPG enzyme. Furthermore, long-term exposure to nano-TiO2 has also proved to induce cell-transformation promoting cell-anchorage independent growth in soft-agar. Results were similar for the two nano-TiO2 sizes. Negative results were obtained when the microparticulated form of TiO2 was tested, indicating the existence of important differences between the microparticulated and nanoparticulated forms. As a conclusion it should be indicated that the observed genotoxic/tranforming effects were only detected at the higher dose tested (1000 µg/mL) what play down the real risk of environmental exposures to this nanomaterial.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
IRMM531A
Titanium, IRMM® certified Reference Material, 0.1 mm foil
Ti
IRMM531B
Titanium, IRMM® certified Reference Material, 0.5 mm foil
Ti
IRMM531C
Titanium, IRMM® certified Reference Material, 0.5 mm wire
Ti
305812
Titanium, crystalline, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Ti
433667
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Ti
369489
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Ti
348805
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Ti
267481
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Ti
267503
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Ti
460397
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Ti
348791
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Ti
348813
Titanium, foil, thickness 0.1 mm, 99.99% trace metals basis
Ti
348821
Titanium, foil, thickness 0.05 mm, 99.99% trace metals basis
Ti
348848
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Ti
268496
Titanium, powder, −100 mesh, 99.7% trace metals basis
Ti
366994
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Ti
513415
Titanium, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 98.5% trace metals basis
Ti
347132
Titanium, rod, diam. 6.35 mm, 99.99% trace metals basis
Ti
268526
Titanium, sponge, 3-19 mm, 99.5% trace metals basis
Ti
348856
Titanium, wire, diam. 2.0 mm, 99.99% trace metals basis
Ti
266035
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Ti
267902
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Ti
348864
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Ti
460400
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Ti
266019
Titanium, wire, diam. 0.127 mm, 99.99% trace metals basis
Ti
749044
Titanium, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.995% trace metals basis
Ti
767506
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Ti
GF57414179
Titanium, rod, 1000mm, diameter 6mm, annealed, 99.6+%
Ti
GF72693883
Titanium, rod, 100mm, diameter 1.5mm, annealed, 99.6+%
Ti
GF76772391
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Ti