EMAIL THIS PAGE TO A FRIEND
Redox biology
Examination of bioenergetic function in the inner mitochondrial membrane peptidase 2-like (Immp2l) mutant mice.
PMID 25460737
Abstract
Inner mitochondrial membrane peptidase 2-like (IMMP2L) protein is a mitochondrial inner membrane peptidase that cleaves the signal peptide sequences of cytochrome c1 (CYC1) and mitochondrial glycerol phosphate dehydrogenase (GPD2). Immp2l mutant mice show infertility and early signs of aging. It is unclear whether mitochondrial respiratory deficiency underlies this phenotype. Here we show that the intermediate forms of GPD2 and CYC1 have normal expression levels and enzymatic function in Immp2l mutants. Mitochondrial respiration is not diminished in isolated mitochondria and cells from mutant mice. Our data suggest that respiratory deficiency is not the cause of the observed Immp2l mutant phenotypes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8674
Antimycin A from Streptomyces sp.