EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Response of aerobic granular sludge to the long-term presence to nanosilver in sequencing batch reactors: reactor performance, sludge property, microbial activity and community.
PMID 25460955
Abstract
The increasing use of silver nanoparticles (Ag NPs) raises concerns about their potential toxic effects on the environment. Granular shape sludge is a special type of microbial aggregate. The response of aerobic granular sludge (AGS) to the long-term presence of Ag NPs has not been well studied. In this study, AGS was exposed to 5 and 50mg/L Ag NPs in sequence batch reactors (SBRs) for 69 days, and its response was evaluated based on the sludge properties, microbial activity and community, and reactor performance. The results showed that Ag NPs caused inhibition to microbial activities of AGS from Day 35. At the end of 69 days of Ag NPs exposure, the microbial activity of AGS was significantly inhibited in terms of inhibitions of the ammonia oxidizing rate (33.0%), respiration rate (17.7% and 45.6%) and denitrification rate (6.8%), as well as decreases in the ammonia mono-oxygenase and nitrate reductase activities. During the long-term exposure, the AGS maintained its granular shape and large granule size (approximately 900 μm); the microbial community of AGS slightly changed, but the dominant microbial population remained. Overall, the AGS tolerated the toxicity of Ag NPs well, but a long-term exposure may produce chronic toxicity to the AGS, which is concerning.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85131
Silver, colloidal, 65-75% Ag basis
Ag
373249
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Ag
327077
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Ag
326976
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Ag
348724
Silver, foil, thickness 1.5 mm, 99.9% trace metals basis
Ag
369438
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Ag
265543
Silver, foil, thickness 1.0 mm, 99.9% trace metals basis
Ag
265535
Silver, foil, thickness 0.5 mm, 99.99% trace metals basis
Ag
345075
Silver, foil, thickness 0.5 mm, 99.9% trace metals basis
Ag
348716
Silver, foil, thickness 0.25 mm, 99.99% trace metals basis
Ag
326984
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Ag
265527
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Ag
326992
Silver, foil, thickness 0.075 mm, 99.9% trace metals basis
Ag
265519
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Ag
303372
Silver, granular, >250 μm, 99.99% trace metals basis
Ag
461032
Silver, needles, 250-600 μm, ≥99.99% trace metals basis
Ag
265500
Silver, powder, <250 μm, 99.99% trace metals basis
Ag
327107
Silver, powder, <45 μm, ≥99.99% trace metals basis
Ag
327093
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Ag
327085
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Ag
484059
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Ag
576832
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Ag
265624
Silver, rod, diam. 7.0 mm, 99.99% trace metals basis
Ag
348775
Silver, rod, diam. 3.2 mm, 99.98% trace metals basis
Ag
204366
Silver, shot, 1-3 mm, 99.999% trace metals basis
Ag
327050
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Ag
267430
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Ag
327018
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Ag
348759
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Ag
265594
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Ag