EMAIL THIS PAGE TO A FRIEND
Cytokine
ADAM17 regulates IL-1 signaling by selectively releasing IL-1 receptor type 2 from the cell surface.
PMID 25461404
Abstract
Interleukin (IL)-1 is one of the most evolutionarily conserved cytokines and plays an essential role in the regulation of innate immunity. IL-1 binds to two different receptors, IL-1R1 and IL-1R2, which share approximately 28% amino acid homology. IL-1R1 contains a cytoplasmic domain and is capable of transducing cellular signals; by contrast, IL-1R2 lacks a functional cytoplasmic domain and serves as a decoy receptor for IL-1. Interestingly, IL-1R2 is proteolytically cleaved and also functions as a soluble receptor that blocks IL-1 activity. In the present study, we examined the shedding properties of IL-1R2 and demonstrate that ADAM17 is de facto the major sheddase for IL-1R2 and that introducing a mutation into the juxta-membrane domain of IL-1R2 significantly desensitizes IL-1R2 to proteolytic cleavage. IL-1R1 was almost insensitive to ADAM17-dependent cleavage; however, the replacement of the juxta-membrane domain of IL-R1 with that of IL-1R2 significantly increased the sensitivity of IL-1R1 to shedding. Furthermore, we demonstrate that ADAM17 indirectly enhances IL-1 signaling in a cell-autonomous manner by selectively cleaving IL-1R2. Taken together, the data collected in the present study indicate that ADAM17 affects sensitivity to IL-1 by changing the balance between IL-1R1 and the decoy receptor IL-1R2.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B8385
Bestatin hydrochloride, ≥98% (HPLC)
C16H24N2O4 · HCl
L8511
Leupeptin, synthetic, ≥85% (HPLC)
C20H38N6O4 · 0.5H2O4S
L5793
Leupeptin, BioUltra, microbial, ≥95% (HPLC)
C20H38N6O4 · 0.5H2O4S
62070
Leupeptin, >4000 U/mg
C20H38N6O4 · 0.5H2O4S