EMAIL THIS PAGE TO A FRIEND
Biochimica et Biophysica Acta
Direct energy transfer from the major antenna to the photosystem II core complexes in the absence of minor antennae in liposomes.
PMID 25461977
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
81780
1,3-Propanediol, puriss., ≥99.0% (GC)
C3H8O2