EMAIL THIS PAGE TO A FRIEND
Photodiagnosis and photodynamic therapy
Photoactivation of curcumin and sodium hypochlorite to enhance antibiofilm efficacy in root canal dentin.
PMID 25462576
Abstract
To test the effect of ultrasonic or light activated curcumin and sodium hypochlorite against Enterococcus faecalis biofilms in vitro. E. faecalis biofilms were grown within root canals (n=175) and divided into 7 groups (n=25). Group 1, sterile saline; group 2, 3% sodium hypochlorite; group 3, 3% sodium hypochlorite activated with ultrasonic files (30s cycles for 4min); group 4, 3% sodium hypochlorite irradiated with blue light (1200mw/cm(2) for 4min); group 5, curcumin (2.5mg/mL); group 6, curcumin (2.5mg/mL) activated with ultrasonic files (30s cycles for 4min); group 7, curcumin (2.5mg/mL) irradiated with blue light. The biofilms' ultrastructure was examined using scanning electron microscopy. Bacterial viability was assessed by confocal microscopy. Data were analyzed by one-way ANOVA and Student-Newman-Keuls test (P=0.05). The quantitative analysis of the colony-forming units was carried out from dentinal shaving and analyzed by One-way ANOVA and Tukey multiple comparison test (P=0.05). All treatment groups showed a significantly higher percentage of dead bacteria than the saline control (P<0.05). The percentage of dead bacteria was significantly higher when light activated curcumin was used (P<0.05). At both depths (200 and 400 microns), light activated curcumin showed no growth of bacteria. Light activation produced significantly higher antibacterial efficacy than ultrasonic agitation, with light activated curcumin producing the maximum elimination of biofilm bacteria within the root canal lumen and dentinal tubules.