EMAIL THIS PAGE TO A FRIEND
Bioorganic chemistry
Synthesis of azobenzenealkylmaleimide probes to photocontrol the enzyme activity of a bacterial histone deacetylase-like amidohydrolase.
PMID 25462992
Abstract
A series of azobenzenealkylmaleimides (AMDs) with different spacer length was synthesized and coupled via Michael-Addition to a specific mutant of a bacterial histone deacetylase-like amidohydrolase (HDAH). Michaelis-Menten parameters (Vmax and Km) were employed to characterize the effect of both, the spacer length and the configuration (cis vs. trans) of the attached azobenzene moiety, on the HDAH enzyme activity. The photoswitch behavior of the AMD/enzyme conjugate activity was clearly influenced by the AMD spacer length. This study highlights the importance of steric rearrangement of the photoswitch with respect to the active site and describes a strategy to optimize the photocontrol of HDAH.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
151173
Ninhydrin, ACS reagent
C9H6O4
N4876
Ninhydrin, suitable for amino acid detection
C9H6O4