EMAIL THIS PAGE TO A FRIEND
Atherosclerosis
GNB3 gene 825 TT variant predicts hard coronary events in the population-based Heinz Nixdorf Recall study.
PMID 25463071
Abstract
The C825T polymorphism of the gene encoding the human G protein beta-3 subunit (GNB3) is associated with hypertension and obesity. Moreover, genotypes of the GNB3 polymorphism have been associated with development of coronary artery disease, and the 825T allele is thought to influence the process of atherosclerosis. However, the potential of the C825T polymorphism to predict coronary events has been poorly explored in a longitudinal setting at the population level. In 4159 Caucasian subjects from the Heinz Nixdorf Recall study cohort (age: 45-75 years, 48% male), genotypes of the GNB3 C825T polymorphism (rs5443) were determined and associated with fatal and non-fatal myocardial infarction (hard coronary events). Established cardiovascular risk factors were used to adjust for confounders. The median follow-up time was 9.9 years (1st/3rd quartiles 9.5/10.2). 148 subjects (3.6%) experienced a hard coronary event. The 10-year event-free survival rate was CC, 96.1%; CT 96.9%, TT, 93.7% (p = 0.018). Multivariable analysis showed that the TT genotype is a significant risk factor for hard coronary events (hazard ratio (HR) = 1.9 (95% confidence interval (CI) 1.2-2.9); p = 0.008) after adjustment for age, sex, diabetes, systolic blood pressure, body mass index, high-density lipoprotein, and coronary artery calcification as determined by electron beam computed tomography at baseline. While prognosis in females was independent of GNB3 genotypes, analysis in males even elevated the HR for TT versus C-allele to 2.6 (95% CI 1.6-4.2; p < 0.001). The GNB3 825 TT genotype is a significant and independent risk factor for hard coronary events independent of other established cardiovascular risk factors at a population level in males.