EMAIL THIS PAGE TO A FRIEND
Journal of Proteomics
SPECHT - single-stage phosphopeptide enrichment and stable-isotope chemical tagging: quantitative phosphoproteomics of insulin action in muscle.
PMID 25463755
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1270000
Arginine, European Pharmacopoeia (EP) Reference Standard
C6H14N4O2
43815
DL-Dithiothreitol, BioUltra, for molecular biology, ≥99.5% (RT)
C4H10O2S2
43819
DL-Dithiothreitol, ≥99.0% (RT)
C4H10O2S2
D0632
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
C4H10O2S2
D5545
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
C4H10O2S2
D9779
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)
C4H10O2S2
D9163
DL-Dithiothreitol, for electrophoresis, ≥99%
C4H10O2S2
58221
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
ClK
03366
Phosphate atomic spectroscopy standard concentrate 1.00 g PO43-, 1.00 g/L, for 1L standard solution, analytical standard
H3O4P
60126
Potassium chloride, tested according to Ph.Eur.
ClK
PHR1329
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClK
60137 Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl
ClK
60135
Potassium chloride solution, BioUltra, ~3 M in H2O
ClK
60134
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
ClK
60136
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
ClK
60138
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
ClK
190020
Sodium cyanoborodeuteride, 96 atom % D, 98% (CP)
CD3BNNa
42077
Sodium cyanoborohydride, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
NaBH3CN
156159
Sodium cyanoborohydride, reagent grade, 95%
NaBH3CN
71435
Sodium cyanoborohydride, purum, ≥95.0% (RT)
NaBH3CN
296813
Sodium cyanoborohydride solution, 1.0 M in THF
CH3BNNa
296945
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
CH3BNNa
243655
Sodium molybdate, ≥98%
MoNa2O4
737860
Sodium molybdate, anhydrous, powder, −100 mesh particle size, 99.9% trace metals basis
MoNa2O4
79299
Sodium tartrate dibasic solution, BioUltra, 1.5 M in H2O (colorless solution at 20 °C)
C4H4Na2O6