EMAIL THIS PAGE TO A FRIEND
Journal of Hazardous Materials
In depth analysis of apoptosis induced by silica coated manganese oxide nanoparticles in vitro.
PMID 25464291
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74738
1-Octadecene, analytical standard
C18H36
74740
1-Octadecene, ≥95.0% (GC)
C18H36
74745
1-Octadecene, technical, ~85% (GC)
C18H36
95446
1-Octanol, analytical standard
C8H18O
74850
1-Octanol, puriss., ≥99.5% (GC)
C8H18O
297887
1-Octanol, anhydrous, ≥99%
C8H18O
W280003
1-Octanol, ≥98%, FCC, FG
C8H18O
W280011
1-Octanol, natural, ≥98%, FCC
C8H18O
650455
Cyclohexane, for HPLC, ≥99.9%
C6H12
34855
Cyclohexane, for HPLC, ≥99.7%
C6H12
33117
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
C6H12
179191
Cyclohexane, ACS reagent, ≥99%
C6H12
676861
Cyclohexane, ACS reagent, ≥99%
C6H12
C100307
Cyclohexane, Laboratory Reagent, ≥99.8%
C6H12
221279
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Cl2Mn · 4H2O
M8054
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Cl2Mn · 4H2O
203734
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Cl2Mn · 4H2O
M3634
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Cl2Mn · 4H2O
M5005
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Cl2Mn · 4H2O
63535
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Cl2Mn · 4H2O
31422
Manganese(II) chloride tetrahydrate, puriss. p.a., ACS reagent, ≥99%
Cl2Mn · 4H2O
O7501
Sodium oleate, ≥99%
C18H33NaO2
O3880
Sodium oleate, ≥95% (capillary GC)
C18H33NaO2
86578
Tetraethyl orthosilicate, ≥99.0% (GC)
C8H20O4Si
131903
Tetraethyl orthosilicate, reagent grade, 98%
C8H20O4Si
759414
Tetraethyl orthosilicate, packaged for use in deposition systems
C8H20O4Si
442272
1-Octadecene, analytical standard
C18H36